Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II Antibody (CVS20) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP234848 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234848 0.1 mg
NB019656 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP234848 Supplier Novus Biologicals Supplier No. NBP234848
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC Class II Monoclonal specifically detects MHC Class II in Human, Bovine, Canine, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen MHC Class II
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone CVS20
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Gene Symbols HLA-DMA
Host Species Mouse
Immunogen 3132 cells
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Asthma, B Cell Development and Differentiation Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3108
Target Species Human, Bovine, Canine, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.