Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II Antibody (ER-TR2) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB100649590.025MG
Description
MHC Class II Monoclonal specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
MHC Class II | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500 | |
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
NB100-64959 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class II molecule. In the mouse, MHC class II molecules are expressed by dendritic cells, B-cells and macrophages. In thymus tissues, clone ER-TR2 stains cortical and medullary stromal cells. Clone ER-TR2 specifically recognizes mouse strains with the haplotypes I-A k,s,r. Mouse strains with the haplotypes I-A b,d,q,f are not recognized by this antibody. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
ER-TR2 | |
Unconjugated | |
Phosphate buffered saline with 0.09% Sodium Azide | |
HLA-DMA | |
Isolated C3H thymic stromal cells | |
0.025 mg | |
Immunology, Major Histocompatibility Complexes | |
3108 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction