Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class II Antibody (ER-TR2) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | MHC Class II |
---|---|
Clone | ER-TR2 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry, Flow Cytometry, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017673
![]() |
Novus Biologicals
NB100649590.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064959
![]() |
Novus Biologicals
NB10064959 |
0.125 mg |
Each for $499.50
|
|
|||||
Description
MHC Class II Monoclonal specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
MHC Class II | |
1.0 mg/mL | |
Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Rat | |
Immunology, Major Histocompatibility Complexes | |
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
HLA-DMA | |
IgG2b | |
Protein G purified | |
NB100-64959 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class II molecule. In the mouse, MHC class II molecules are expressed by dendritic cells, B-cells and macrophages. In thymus tissues, clone ER-TR2 stains cortical and medullary stromal cells. Clone ER-TR2 specifically recognizes mouse strains with the haplotypes I-A k,s,r. Mouse strains with the haplotypes I-A b,d,q,f are not recognized by this antibody. |
ER-TR2 | |
Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
3108 | |
Isolated C3H thymic stromal cells | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title