Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II Antibody (ER-TR2), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB209959 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB209959 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB209959 Supplier Novus Biologicals Supplier No. NB10064959DL594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MHC Class II Monoclonal antibody specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)

Specifications

Antigen MHC Class II
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ER-TR2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Host Species Rat
Immunogen Isolated C3H thymic stromal cells
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3108
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.