Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II Antibody (HIS19), FITC, Novus Biologicals™
SDP

Catalog No. NBP143985 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143985 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143985 Supplier Novus Biologicals Supplier No. NBP143985

Mouse Monoclonal Antibody

MHC Class II Monoclonal specifically detects MHC Class II in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen MHC Class II
Applications Flow Cytometry
Classification Monoclonal
Clone HIS19
Concentration 0.5 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1:10-1:1000
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Gene Symbols HLA-DMA
Host Species Mouse
Immunogen The immunogen for this antibody is MHC Class II.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immune System Diseases, Immunology, Major Histocompatibility Complexes, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3108
Target Species Rat
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.