Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II Antibody (NIMR-4), FITC, Novus Biologicals™
SDP

Catalog No. NBP128159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128159 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128159 Supplier Novus Biologicals Supplier No. NBP128159
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MHC Class II Monoclonal specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.

Specifications

Antigen MHC Class II
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone NIMR-4
Concentration 0.5mg/mL
Conjugate FITC
Dilution Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Gene Symbols HLA-DMA
Host Species Rat
Immunogen The immunogen is a peptide made to the amino acid region MHC Class II
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3108
Test Specificity MHC Class II molecules
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.