Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II antigen I Ak,d,b,q,r Antibody (ER-TR3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB10064961J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064961J 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064961J Supplier Novus Biologicals Supplier No. NB10064961APC
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MHC Class II antigen I Ak,d,b,q,r Monoclonal specifically detects MHC Class II antigen I Ak,d,b,q,r in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen MHC Class II antigen I Ak,d,b,q,r
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ER-TR3
Conjugate APC
Dilution Flow Cytometry
Gene Alias Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly
Gene Symbols HLA-DRB1
Host Species Rat
Immunogen Isolated C3H thymic stromal cells
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3123
Test Specificity NB100-64961 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class II molecule. In the mouse, MHC class II molecules are expressed by dendritic cells, B-cells and macrophages. In thymus tissues, clone ER-TR3 stains cortical and medullary stromal cells. Clone ER-TR3 specifically recognizes mouse strains with the haplotypes I-A k,b,d,q,r. Mouse strains with the haplotypes I-A s,f are not recognized by this antibody.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.