Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC class II (I-A/I-E) Antibody (M5/114.15.2), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N143312APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N143312APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N143312APC7 Supplier Novus Biologicals Supplier No. NBP143312APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MHC class II (I-A/I-E) Monoclonal specifically detects MHC class II (I-A/I-E) in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen MHC class II (I-A/I-E)
Applications Flow Cytometry
Classification Monoclonal
Clone M5/114.15.2
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Gene Symbols H2-Ea-ps
Host Species Rat
Immunogen The immunogen for this antibody was MHC Class II.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Major Histocompatibility Complexes, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 100504404
Test Specificity The monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.