Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC class II (I-A/I-E) Antibody (MK-D6), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB384023 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384023 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384023 Supplier Novus Biologicals Supplier No. NBP308856R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC class II (I-A/I-E) Monoclonal antibody specifically detects MHC class II (I-A/I-E) in Mouse, Human (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MHC class II (I-A/I-E)
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone MK-D6
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias E-alpha-f;H-2 class II histocompatibility antigen, E-K alpha chain;H-2 class II histocompatibility antigen, E-U alpha chain;H-2E;H2-Ea;H-2Ea;H2-Ea-ps;I;Ia;Ia3;Ia-3;I-E;I-Ealpha;MHC-H2-Ea
Host Species Mouse
Immunogen Spleen cells from B10.D2 mice
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 100504404
Target Species Mouse, Human (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.