Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II RT1B Antibody (OX-6), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB339439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339439 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339439 Supplier Novus Biologicals Supplier No. NB10065541AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC Class II RT1B Monoclonal antibody specifically detects MHC Class II RT1B in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating, CyTOF

Specifications

Antigen MHC Class II RT1B
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OX-6
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DMA, DMB, DP beta 1, DPB 1, DPB1, DRB, HLA class II histocompatibility antigen DM beta chain, HLA DMB, HLA DPB1, HLADM, HLADP1B, Major histocompatibility complex class II, Major histocompatibility complex class II DP beta 1, MHC class II antigen DMB, MHC DPB1, RING6, RING7
Host Species Mouse
Immunogen Rat thymocyte membrane glycoproteins
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 24738
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.