Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II RT1Bu Antibody (OX-3), PE, Novus Biologicals™
SDP

Catalog No. NB10065011Q Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065011Q 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065011Q Supplier Novus Biologicals Supplier No. NB10065011PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC Class II RT1Bu Monoclonal specifically detects MHC Class II RT1Bu in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen MHC Class II RT1Bu
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OX-3
Conjugate PE
Dilution Flow Cytometry
Gene Alias Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly
Gene Symbols HLA-DRB1
Host Species Mouse
Immunogen Rat thymocyte membrane glycoproteins
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3123
Test Specificity NB100-65011 recognizes a polymorphic determinant of rat Ia antigen (RT1Bu) present on Lewis, Wistar and AO strain rats but not BN, DA or PVG/c strains (1,2). This antibody is useful for distinguishing Ia positive cells from different rat strains, e.g. for recognizing cells of donor origin in bone marrow reconstituted radiation chimaeras (3,4). MRC OX-3 cross reacts with mouse strains of MHC haplotypes b and s, and analysis of recombinant mouse strains showed that the determinants mapped to the I-A region. NB100-65011 recognizes Ia antigens on B-cells, dendritic cells and certain epithelial cells (1,3,4). This product is routinely tested in flow cytometry on Lewis rat splenocytes.
Target Species Rat
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.