Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIA Antibody, Novus Biologicals™
SDP

Catalog No. NBP226185 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226185 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226185 Supplier Novus Biologicals Supplier No. NBP226185
Add to Cart
Add to Cart

Goat Polyclonal Antibody

MIA Polyclonal specifically detects MIA in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen MIA
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias CD-RAP, melanoma inhibitory activity, Melanoma inhibitory activity protein, melanoma-derived growth regulatory protein
Gene Symbols MIA
Host Species Goat
Immunogen Peptide with sequence PKLADRKLCADQE corresponding to internal region according to NP_006524.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8190
Test Specificity The immunizing peptide represents the N terminus of the mature protein. Reported variants represent identical protein: NP_006524.1, NP_001189482.1,
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.