Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIA3 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB104860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104860 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104860 Supplier Novus Biologicals Supplier No. NBP298635R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MIA3 Polyclonal specifically detects MIA3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MIA3
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias ARNT, C219 Reactive Peptide, C219-Reactive Peptide, D320, KIAA0268, Melanoma Inhibitory Activity Family, Member 3, Melanoma Inhibitory Activity Protein 3, TANGO, TANGO1, Transport And Golgi Organization, Transport And Golgi Organization Protein 1, UNQ6077
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MIA3 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375056
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.