Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119286
This item is not returnable.
View return policy
Description
MIB2 Polyclonal Antibody for Western Blot, IHC (P)
E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. Positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors.
Specifications
MIB2 | |
Polyclonal | |
Unconjugated | |
MIB2 | |
2210008I11Rik; Dyslike; dystrophin-like protein; E3 ubiquitin-protein ligase MIB2; EC 6.3.2.13; FLJ20648; FLJ41406; Mib2; Mind bomb homolog 2; mind bomb-2; mindbomb E3 ubiquitin protein ligase 2; mindbomb homolog 2; mindbomb homolog 2 (Drosophila); novel zinc finger protein; Novelzin; putative NF-kappa-B-activating protein 002N; RBSC-skeletrophin; RBSC-skeletrophin/dystrophin-like polypeptide; RING-type E3 ubiquitin transferase MIB2; skd; Skeletrophin; zinc finger ZZ type with ankyrin repeat domain protein 1; zinc finger, ZZ type with ankyrin repeat domain 1; Zzank1; ZZZ5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
474147 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q68LP1 | |
MIB2 | |
Recombinant protein Mindbomb Homolog 2. The antigen corresponds to amino acid range 407-663 of the target protein. | |
200 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction