Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $577.00
Specifications
Antigen | MIB2 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119286
|
Invitrogen™
PA5119286 |
200 μL |
Each for $577.00
|
|
|||||
PIPA5121773
|
Invitrogen™
PA5121773 |
20 μL |
Each for $237.00
|
|
|||||
Description
MIB2 Polyclonal Antibody for Western Blot, IHC (P)
E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. Positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors.Specifications
MIB2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Rat | |
MIB2 | |
2210008I11Rik; Dyslike; dystrophin-like protein; E3 ubiquitin-protein ligase MIB2; EC 6.3.2.13; FLJ20648; FLJ41406; Mib2; Mind bomb homolog 2; mind bomb-2; mindbomb E3 ubiquitin protein ligase 2; mindbomb homolog 2; mindbomb homolog 2 (Drosophila); novel zinc finger protein; Novelzin; putative NF-kappa-B-activating protein 002N; RBSC-skeletrophin; RBSC-skeletrophin/dystrophin-like polypeptide; RING-type E3 ubiquitin transferase MIB2; skd; Skeletrophin; zinc finger ZZ type with ankyrin repeat domain protein 1; zinc finger, ZZ type with ankyrin repeat domain 1; Zzank1; ZZZ5 | |
MIB2 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q68LP1 | |
474147 | |
Recombinant protein Mindbomb Homolog 2. The antigen corresponds to amino acid range 407-663 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title