Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIB2/Skeletrophin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$155.13
Specifications
| Antigen | MIB2/Skeletrophin |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-0328
|
Bethyl Laboratories, Inc
A301414AT |
20 μL |
Each of 1 for $155.13
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. Positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors.Specifications
| MIB2/Skeletrophin | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| MIB2, Mind Bomb-2, Novelzin, SKD, Skeletrophin, ZZANK1, ZZZ5, E3 ubiquitin-protein ligase MIB2, EC 6.3.2, EC 6.3.2.-, EC 6.3.2.13, FLJ20648, FLJ41406, Mind bomb homolog 2, mindbomb homolog 2 (Drosophila), Novel zinc finger protein, novelzin, Novelzin, Putative NF-kappa-B-activating protein 002N, SKD, Skeletrophin, Zinc finger ZZ type with ankyrin repeat domain protein 1, ZZ type with ankyrin repeat domain 1, ZZANK1 | |
| MIB2 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96AX9 | |
| 142678 | |
| Between 925 and 975 | |
| Primary | |
| 4° C | |
| MIB2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title