Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICA Antibody (159227), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB350024 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350024 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350024 Supplier Novus Biologicals Supplier No. FAB1300MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MICA Monoclonal specifically detects MICA in Human samples. It is validated for Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry, Gene Knock-Out, Western Blot.

Specifications

Antigen MICA
Applications Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry, Gene Knock-Out, Western Blot
Classification Monoclonal
Clone 159227
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias FLJ60820, MGC111087, PERB11.1
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human MICA
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 100507436
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.