Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICA/B Antibody (159207), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314629 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314629 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314629 Supplier Novus Biologicals Supplier No. FAB13001PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MICA/B Monoclonal specifically detects MICA/B in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MICA/B
Applications Flow Cytometry
Classification Monoclonal
Clone 159207
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human MICA, Accession # Q29983
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100507436
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.