Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICA/B Antibody (6D4), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB163398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163398 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB163398 Supplier Novus Biologicals Supplier No. NBP276811C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MICA/B Monoclonal antibody specifically detects MICA/B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry

Specifications

Antigen MICA/B
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 6D4
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2
Host Species Mouse
Immunogen Transfected C1R cells expressing MICA
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100507436
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2 α
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.