Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICALL1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB163368 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163368 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163368 Supplier Novus Biologicals Supplier No. NBP270017APC
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MICALL1 Polyclonal antibody specifically detects MICALL1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen MICALL1
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias DKFZp686M2226, KIAA1668, MICAL-L1, MICAL-like 1, MICAL-like protein 1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human MICALL1 protein (between amino acids 200-280) [UniProt Q8N3F8]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 85377
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.