Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICB Antibody (102), PE, Novus Biologicals™
SDP

Catalog No. NB306789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306789 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306789 Supplier Novus Biologicals Supplier No. NBP289627PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for Western Blot

Specifications

Antigen MICB
Applications Western Blot
Classification Monoclonal
Clone 102
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human MICB (Accession#: NP_005922.2; Met1-Gly298).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 4277
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.