Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICB Antibody (MM0473-3C37), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP211798Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211798Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211798Y Supplier Novus Biologicals Supplier No. NBP211798AF647

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

MICB Monoclonal specifically detects MICB in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen MICB
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MM0473-3C37
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry
Gene Accession No. NP_001170990
Gene Alias MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog
Gene Symbols MICB
Host Species Mouse
Immunogen Human recombinant MICB
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4277
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.