Learn More
MilliporeSigma™ Supelco™ Michel-Miller Column Plug Front Seal, PTFE
Catalog No. p-7191296 Shop All MilliporeSigma Supelco Products
Click to view available options
Thread Size:
80 mm
Solid plug for use as stopper with Ace-Threds. Seals M-M column ends, Plugs with No. 50 Ace-Thred fit Ace large columns.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Thread Size 80 mm
Material PTFE
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.