Learn More
MilliporeSigma™ Supelco™ Michel-Miller Swagelok™ adapter NPTF 1/8 in., Nylon
Catalog No. p-7191384 Shop All MilliporeSigma Supelco Products
Click to view available options
Thread Size:
Ace-Thred No. 15
Ace-Thred No. 25
Use to make Swagelok™ tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with No. 50 Ace-Thred fit Ace large columns.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Thread Size Ace-Thred No. 25
Material Nylon
Swagelok is a registered trademark of Swagelok Company
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.