Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICL/CLEC12A Antibody (2439B), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB363641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363641 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363641 Supplier Novus Biologicals Supplier No. FAB29461W
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MICL/CLEC12A Monoclonal antibody specifically detects MICL/CLEC12A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF

Specifications

Antigen MICL/CLEC12A
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 2439B
Conjugate DyLight 650
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A, Thr67-Ala265, Accession # Q5QGZ9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160364
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.