Learn More
Description
Specifications
Specifications
| Antigen | MICL/CLEC12A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2439B |
| Conjugate | PE-Cyanine5.5 |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL |
| Host Species | Rabbit |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A, Thr67-Ala265, Accession # Q5QGZ9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
