Learn More
3M™ Microbial Luminescence System Injector Cleaning Kit 3005 DPQCOG-CLNKIT, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. s27265 Shop All AquaPhoenix Scientific Products
Click to view available options
:
Microbial Luminescence System Injector Cleaning Kit 3005 DPQCOG-CLNKIT, Sold by AquaPhoenix Scientific
Prevents biofilm build-up
  • For daily use
  • Store 2 to 8°C
  • Includes:
    • 2 cleaning solutions (125 mL each)
    • 10 ATP-free water (125 mL each)
    • 10 amber vials
Specifications
Product Type Cleaning Kit
Includes 10 ATP-free water, 2 clean solutions and 10 amber vials
For Use With (Application) Preventing biofilm build-up
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.