Learn More
Microbiologics™ Epower™ Aspergillus brasiliensis ATCC™ 16404™†
SDP
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
$1039.00
Specifications
Biosafety Level 1
Form Lyophilized Pellet
Quantity 10/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
12-111-235
SDP
View Documents
Microbiologics™
0392E6CRM
1.0-9.9 + 6 CFU/pellet
Each for $1,039.00
Only null left
Add to Cart
12-111-234
SDP
View Documents
Microbiologics™
0392E3CRM
1.0-9.9 + 3 CFU/pellet
Each for $1,039.00
Only null left
Add to Cart
Description
Description
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 16404™†
Specifications
Specifications
1
Lyophilized Pellet
10/Pk.
Videos
SDS
Documents
Documents
Product Certifications
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.