Learn More
Microbiologics™ Epower™ Enterococcus faecalis, ATCC™ 29212™†
Shop All Microbiologics Products

Description
Includes
Product warranty and peel-off certificate of assay
Microorganism preparations manufactured at a predetermined concentration for versatile laboratory testing requirements.
- Process controls from one hundred to over a billion CFU/preparation
- Shelf life of 10-20 months Traceable to reference cultures
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Specifications
Specifications
| ATCC® | 29212 |
| Biosafety Level | 2 |
| Form | Lyophilized Pellet |
| Product Type | QC Microorganism |
| For Use With (Application) | Lyophilized pellets add convenience to special projects and internal proficiency plans. |
| Includes | Product warranty and peel-off certificate of assay |
| Mean Assay Value | 1.0-9.9E+03 CFU per pellet |
| Packaging Type | Vial |
| Quantity | 10/Pk. |
| Species | Enterococcus faecalis |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.