Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Epower™ Salmonella enterica subsp. enterica serovar Typhimurium, ATCC™ 14028™†
Shop All Microbiologics Products

Click to view available options
Mean Assay Value:
1.0-9.9E+03 CFU per pellet
1.0-9.9E+04 CFU per pellet
Description
Includes
Product warranty and peel-off certificate of assay
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Specifications
Specifications
ATCC® | 14028 |
Biosafety Level | 2 |
Form | Lyophilized Pellet |
Product Type | QC Microorganism |
For Use With (Application) | Lyophilized pellets add convenience to special projects and internal proficiency plans. |
Includes | Product warranty and peel-off certificate of assay |
Mean Assay Value | 1.0-9.9E+03 CFU per pellet |
Packaging Type | Vial |
Quantity | 10/Pk. |
Species | Salmonella enterica subsp. enterica serovar typhimurium |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction