Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ EZ-FPC™ Microorganism Preparations
Shop All Microbiologics Products

Click to view available options
Species:
Alicyclobacillus acidoterrestris
Escherichia coli
Lactobacillus rhamnosus
ATCC®:
ATCC 43888
ATCC 49025
ATCC 9595
Description
Includes
Product warranty and peel-off Certificate of Assay
- Available in 100 to 1000CFU per preparation (qualitative) or 1000 to 10,000CFU per preparation (quantitative)
- Daily process controls for food quality labs
- Traceable to reference cultures
Order Info
Packaged in a vial of a single assayed microorganism (10 lyophilized pellets in each vial), with an attached Certificate of Assay
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications
Specifications
ATCC® | ATCC 43888 |
Format | EZ-FPC |
Product Type | EZ-FPC |
For Use With (Application) | Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls. |
Includes | Product warranty and peel-off certificate of assay |
Packaging Type | Vial |
Species | Escherichia coli |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction