Learn More
Microbiologics™ EZ-PEC™ Microorganism Preparations
Shop All Microbiologics Products

Description
Includes
Product warranty and peel-off Certificate of Assay
Predetermined mean assay value results in a final challenge concentration of 100,000 to 1,000,000CFU/1.0mL of product tested
- Enumerated lyophilized microorganism preparations are designed to provide the required microorganism challenge concentrations for Antimicrobial Effectiveness and Preservative Efficacy Tests
- Traceable to reference cultures
Order Info
Each kit contains 20 lyophilized pellets of a single enumerated strain plus ten vials of hydrating fluid (2mL each)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications
Specifications
| ATCC® | NCIMB 8545™* |
| Biosafety Level | 2 |
| Format | EZ-PEC |
| For Use With (Application) | Add convenience to pharmaceutical and cosmetic testing labs needing to meet antimicrobial effectiveness or preservative efficacy testing. |
| Includes | Product warranty and peel-off certificate of assay |
| Packaging Type | Vial |
| Quantity | 2 Vials of 10 Pellets |
| Species | Escherichia coli |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.