Learn More
Microbiologics™ Clostridium difficile ATCC™ BAA-1870™†
SDP
For culture purposes and QC applications
Specifications
Biosafety Level 2
Species Clostridium difficile
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
23-002-132
SDP
View Documents
Microbiologics™
01048P
KWIK-STIK 2/Pk.
Pack of 2 for $152.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-016-022
SDP
View Documents
Microbiologics™
01048K
KWIK-STIK 6/Pk.
Pack of 6 for $393.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-016-458
SDP
View Documents
Microbiologics™
01048L
Lyfo-Disk QC Control 6/Pk.
Pack of 6 for $368.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Clostridium difficile strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Clostridium difficile strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ BAA-1870™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
Specifications
2
Clostridium difficile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.