Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single Corynebacterium renale strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2 or 6
- The most economical reference stock culture option; lyophilized pellet containing a single Corynebacterium renale strain
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing 6 pellets
- ATCC™ BAA-1785™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
Specifications
| ATCC® | BAA-1785 |
| Biosafety Level | 2 |
| Product Type | Lyfo-Disk QC Control |
| Quantity | 6/Pk. |
| Species | Corynebacterium renale |