Learn More
Microbiologics™ Eggerthella lenta ATCC™ 43055™†
SDP
For culture purposes and QC applications
Supplier:  Microbiologics™ 0936L
 View more versions of this product
Catalog No. 23-016-889
Only null left
Add to Cart
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Eggerthella lenta strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Eggerthella lenta strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 43055™†
Specifications
Specifications
43055
1
LYFO DISK™
6/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.