Learn More
Microbiologics™ Eggerthella lenta ATCC™ 43055™†
SDP
For culture purposes and QC applications
$206.00 - $344.00
Specifications
ATCC® 43055
Biosafety Level 1
View More Specs
Products 2
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
23-021267
SDP
View Documents
Microbiologics™
0936P
KWIK-STIK™
Pack of 2 for $206.00
Only null left
Add to Cart
23-016-889
SDP
View Documents
Microbiologics™
0936L
LYFO DISK™
Pack of 6 for $344.00
Only null left
Add to Cart
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Eggerthella lenta strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Eggerthella lenta strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 43055™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
Specifications
43055
1
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.