Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Scientific™ Microcentrifuge Tubes

Catalog No. p-313988
Click to view available options
Color:
Blue
Green
Yellow

Specifications

Certifications/Compliance RNase, DNase, Endotoxin-free
Autoclavable Autoclavable
Capacity (Metric) 1.7 mL
Color Green
Material Polypropylene
For Use With (Application) Ideal for custom gene libraries, protein work, cDNA synthesis, and nucleic acid amplification
Graduated Yes
Max. RCF 17,000 x g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.