Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell™ MICROCHEM™ by AlphaTec™ Overboots with Multi-Layer Chemical Barrier
Shop All Ansell Products
Click to view available options
Size:
12 to 14
8 to 12
Description
- Durable
- Highly visible bright yellow color for improved worker safety
- Lightweight and durable
- Applications:
- Unexpected leakages spills or other releases
- Loading and unloading trucks and vehicles
- Product inspection
- Cleaning of plant and machines

Specifications
Specifications
Certifications/Compliance | CE CATEGORY I |
Closure Type | Elastic to top of boot with tie fastenings |
Color | Yellow |
Material | Multi-layer Non-woven Barrier Laminate |
Seam Type | Welded |
Size | 8 to 12 |
Packaging Quantity | 300/case |
Resistant To | Liquid, Particles |
Product Type | Overboot |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction