Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Microcline Feldspar Mineral Specimen
SCI_ED

Catalog No. p-7076825
Click to view available options
Quantity:
1/Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Microcline Feldspar Mineral Specimen for use in study of physical properties like hardness, cleavage, and crystallization temperature.

  • For use in classroom identification labs and exercises
  • Hardness of 6 on Mohs scale
  • Distinctly shows two planes of cleavage
  • Differing colors among samples offer visual demonstration of chemical variation
  • Correlates well to Next Generation Science Standards Evidence Statements

Specifications

Product Type Mineral, Microcline Feldspar
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.