Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microcrystalline Cellulose, NF, Spectrum™ Chemical
SDP

Catalog No. 18604592 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-592 500 g Poly Bottle
18-604-593 2.5 kg Poly Pail
18-604-595 12 kg Fiber Drum
18-604-594 50 kg Fiber Drum
18-604-591 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-592 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1679500GM
Only null left
Add to Cart
Add to Cart

C1679, 9004-34-6

Spectrum™ Chemical Microcrystalline Cellulose, NF is an excipient in pharmaceutical formulations acting as a binder in tablet formation. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-34-6
Molecular Formula C14H26O11
Molecular Weight (g/mol) 370.35
InChI Key PTHCMJGKKRQCBF-UHFFFAOYNA-N
IUPAC Name 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
SMILES COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O

Specifications

CAS Max % 100%
Quantity 500 g
Grade NF
Ignition Residue 0.1%
Loss on Drying 7%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.