Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Microliter Sample Syringes - Gas Phase
GREENER_CHOICESDP

Catalog No. p-7221839 Shop All Kemtech Products
Click to view available options
Volume (Metric):
0.5 μL
1 μL
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1000 μL

Catalog No. 31-502-301

Supplier: Synthware™ S371001

Only null left
Add to Cart
Add to Cart

Borosilicate syringes with a beveled tip, for precise sample delivery in volumes ranging from 0.5 μL to 1 mL.

  • Borosilicate glass syringes with a beveled tip
  • For precise sample delivery
  • Gas phase

Specifications

Material Borosilicate Glass (3.3)
Color Clear
For Use With (Application) Precise Sampling
Latex Free Yes
Product Type Syringe
Disposable Reusable
Volume (Metric) 0.5 μL
Needle Point Style Bevel
For Use With (Equipment) Syringe Needles
Connection Type Slip Tip
Graduated Yes
Sterility Non-sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.