Learn More
Restek™ Micropacked Inlet Conversion Kits
SDP
Catalog No. 06611209 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Micropacked Column FID Adaptor Kit
Micropacked Column FID and Injection Port Adaptor Kit
Micropacked Column Injection Port Adaptor Kit
For Use With (Application):
FID
On-Column Injection
Split/Splitless Injection
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Application)
06-611-209 Micropacked Column FID and Injection Port Adaptor Kit Split/Splitless Injection
06-611-213 Micropacked Column FID Adaptor Kit FID
06-611-212 Micropacked Column Injection Port Adaptor Kit On-Column Injection
06-611-211 Micropacked Column Injection Port Adaptor Kit Split/Splitless Injection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-611-209 Supplier Restek™ Supplier No. 22424
Add to Cart
Add to Cart
Convert a Capillary GC Split/Splitless Inlet for use with 1/16 in. O.D. Micropacked Columns
  • For use with Agilent 5890, 6890, and 7890 GCs
  • Sample pathways deactivated for ultimate inertness
Specifications
For Use With (Application) Split/Splitless Injection
Includes Dual Vespel Ring Inlet Seal (Large Bore), Reducing Nut (Large Bore), FID Adaptor (Large Bore), 1/4 in. Ferrule (Vespel/Graphite), 1/4 in. Nut (Stainless Steel), Two 1/16 in. Ferrules (Vespel/Graphite), 4 mm Splitless Liner (Standard Deactivation); Two 1/16 in. Nuts (Stainless Steel)
Type Micropacked Column FID and Injection Port Adaptor Kit
For Use With (Equipment) Packed Column FIDs Only
Functionality/Interaction Type Sample pathways deactivated for ultimate inertness