Description
- For use with Agilent 5890, 6890, and 7890 GCs
- Sample pathways deactivated for ultimate inertness
Specifications
Specifications
| For Use With (Application) | Split/Splitless Injection |
| Includes | Dual Vespel Ring Inlet Seal (Large Bore), Reducing Nut (Large Bore), 1/16 in. Ferrule (Vespel/Graphite), 1/16 in. Nut (Stainless Steel), 4 mm Splitless Liner (Standard Deactivation) |
| Type | Micropacked Column Injection Port Adaptor Kit |
| Functionality/Interaction Type | Sample pathways deactivated for ultimate inertness |