Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Protein LoBind™ Deepwell Plates

Catalog No. 05414247
Click to view available options
Color:
White
White, Clear Bottom
Yellow
No. of Wells:
96
384
Volume (Metric) Well:
140 μL
200 μL
500 μL
1000 μL
2000 μL
Packaging:
10 Bags x 12 Plates
10 Bags x 24 Plates
10 Bags x 8 Plates
5 Bags x 4 Plates
5 Bags x 8 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.

  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing in plates.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Packaging 10 Bags x 24 Plates
Color White
Product Type Protein LoBind™ Plate
Material Polypropylene
Volume (Metric) Well 140 μL
No. of Wells 384
Sterility Non-sterile
Total Volume (Metric) Well 140 μL
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 240 Plates
Well Color Colorless
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.