Learn More
United Scientific™ Microscope Slide Mailer
SDPSCI_ED
Catalog No. p-200009359 Shop All United Scientific Supplies, Inc Products
Click to view available options
Slide Capacity:
1
2
5
10
Catalog No. S15743
Supplier: United Scientific™ 62100
Only null left
Add to Cart
Add to Cart
Allows for the safe storage and transport of microscope slides.
  • These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
  • Available in four sizes.
  • These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
  • Available in four sizes.
Specifications
Type Slide Mailer
Slide Capacity 1
For Use With (Equipment) Microscope Slides
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.