Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MID1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP126612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP126612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP126612 Supplier Novus Biologicals Supplier No. NBP126612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MID1 Polyclonal specifically detects MID1 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).

Specifications

Antigen MID1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_000372.1
Gene Alias EC 6.3.2.-, FLJ57031, FLJ58683, FLJ76288, FXYGBBB1, Midin, midline 1 (Opitz/BBB syndrome), RNF59MIDIN, TRIM18mouse, homolog of
Gene Symbols MID1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 617 and 667 of human midline 1 using the numbering given in entry NP_000372.1 (GeneID 4281).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 4281
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Duckbill platypus, White-tufted-ear marmoset, Crab-eating macaque, African elephant and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.