Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MID1IP1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB063935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB063935 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB063935 Supplier Novus Biologicals Supplier No. NBP300003H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MID1IP1 Polyclonal specifically detects MID1IP1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MID1IP1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias FLJ10386, G12-like, Gastrulation-specific G12-like protein, MID1 interacting protein 1 (gastrulation specific G12 homolog (zebrafish)), MID1 interacting protein 1 (gastrulation specific G12-like (zebrafish)), MID1 interacting protein 1 (gastrulation specific G12-like), Mid1-interacting G12-like protein, mid1-interacting protein 1, MIG12MID1 interacting G12-like protein, Protein STRAIT11499, S14R, Spot 14-R, Spot 14-related protein, STRAIT11499, THRSPL
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant MID1IP1 (Q9NPA3; Met1-His183).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58526
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.