Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MID2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001167 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001167 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001167 Supplier Novus Biologicals Supplier No. NB1001167
Add to Cart
Add to Cart

Goat Polyclonal Antibody

MID2 Polyclonal specifically detects MID2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen MID2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_036348
Gene Alias EC 6.3.2.-, FLJ37715, FLJ41813, FXY2midin-2, midin 2, Midin-2, midline 2, Midline defect 2, Midline-2, RING finger protein 60, RNF60midline-2, TRIM1probable E3 ubiquitin-protein ligase MID2, tripartite motif protein 1, Tripartite motif-containing protein 1
Gene Symbols MID2
Host Species Goat
Immunogen Peptide with sequence QESPYVSGMKTCH corresponding to C-Terminus according to NP_036348.2, NP_438112.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 11043
Test Specificity This antibody is expected to recognize both human isoforms of this protein.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.