Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

midA Antibody (OTI4A5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB287892 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287892 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287892 Supplier Novus Biologicals Supplier No. NBP272681APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

midA Monoclonal antibody specifically detects midA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen midA
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4A5
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias chromosome 2 open reading frame 56, MidA, mitochondrial, PRO1853
Host Species Mouse
Immunogen Full length human recombinant protein of human C2orf56(NP_653337) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55471
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.