Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIF Rabbit anti-Human, Clone: JM11-64, Novus Biologicals™
SDP

Catalog No. NBP266949 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266949 Supplier Novus Biologicals Supplier No. NBP266949

Rabbit Monoclonal Antibody

MIF Monoclonal specifically detects MIF in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen MIF
Applications ChIP Assay, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone JM11-64
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10 - 1:50, Immunoprecipitation 1:10 - 1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase
Gene Symbols MIF
Host Species Rabbit
Immunogen Synthetic peptide within human MIF aa 60-90. (SwissProt: P14174 Human; SwissProt: P34884 Mouse; SwissProt: P30904 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 4282
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.